Imprimer

Anti-SNAP25 Mouse monoclonal antibody [clone: SP12]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24732-100EA 540 EUR
ABCAAB24732-100
Anti-SNAP25 Mouse monoclonal antibody [clone: SP12]
Type: Primary
Antigen: SNAP25 (synaptosomal-associated protein, 25kDa)
Clonality: monoclonal
Clone: SP12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR