Imprimer

Anti-C1QBP Mouse monoclonal antibody [clone: 74.5.2]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24734-100EA 579 EUR
ABCAAB24734-100
Anti-C1QBP Mouse monoclonal antibody [clone: 74.5.2]
Type: Primary
Antigen: C1QBP (complement component 1, q subcomponent binding protein)
Clonality: monoclonal
Clone: 74.5.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR