Imprimer

Anti-P180 Lamellar Body Protein Mouse Monoclonal Antibody [clone: 3C9]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24751-100EA 918 EUR
ABCAAB24751-100
Anti-P180 Lamellar Body Protein Mouse Monoclonal Antibody [clone: 3C9]
Anticorps
Type: Primary
Antigen: P180 Lamellar Body Protein
Clonality: monoclonal
Clone: 3C9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR