Imprimer

Anti-Parathyroid Hormone Receptor 1 Mouse monoclonal antibody [clone: OK1]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24752-100EA 550 EUR
ABCAAB24752-100
Anti-Parathyroid Hormone Receptor 1 Mouse monoclonal antibody [clone: OK1]
Type: Primary
Antigen: Parathyroid Hormone Receptor 1
Clonality: monoclonal
Clone: OK1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR