Imprimer

Anti-AKT1 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 2H12]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24818-100EA 572 EUR
ABCAAB24818-100
Anti-AKT1 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 2H12]
Type: Primary
Antigen: AKT1 (v-akt murine thymoma viral oncogene homolog 1)
Clonality: monoclonal
Clone: 2H12
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR