Imprimer

Anti-GRSF1 Rabbit Polyclonal Antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24830-50EA 674 EUR
ABCAAB24830-50
Anti-GRSF1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: GRSF1 (G-rich RNA sequence binding factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR