Imprimer

Anti-ICAM1 Rat monoclonal antibody FITC (Fluorescein) [clone: YN1/1.7.4]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24869-50EA 298 EUR
ABCAAB24869-50
Anti-ICAM1 Rat monoclonal antibody FITC (Fluorescein) [clone: YN1/1.7.4]
Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: monoclonal
Clone: YN1/1.7.4
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR