Imprimer

Anti-CD45R Rat monoclonal antibody FITC (Fluorescein) [clone: RA3-6B2]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24897-50EA 619 EUR
ABCAAB24897-50
Anti-CD45R Rat monoclonal antibody FITC (Fluorescein) [clone: RA3-6B2]
Type: Primary
Antigen: CD45R (CD45R)
Clonality: monoclonal
Clone: RA3-6B2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR