Imprimer

Anti-EGFR Mouse monoclonal antibody [clone: 9H2]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24912-50EA 586 EUR
ABCAAB24912-50
Anti-EGFR Mouse monoclonal antibody [clone: 9H2]
Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: monoclonal
Clone: 9H2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR