Imprimer

Anti-CTNNB1 Mouse monoclonal antibody [clone: 24E1]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24913-50EA 621 EUR
ABCAAB24913-50
Anti-CTNNB1 Mouse monoclonal antibody [clone: 24E1]
Type: Primary
Antigen: CTNNB1 (catenin (cadherin-associated protein), beta 1, 88kDa)
Clonality: monoclonal
Clone: 240
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR