Imprimer

Anti-CD45.1 Mouse monoclonal antibody FITC (Fluorescein) [clone: A20]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24917-50EA 399 EUR
ABCAAB24917-50
Anti-CD45.1 Mouse monoclonal antibody FITC (Fluorescein) [clone: A20]
Type: Primary
Antigen: CD45.1 (CD45.1)
Clonality: monoclonal
Clone: A20
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR