Imprimer

Anti-EGFR Mouse monoclonal antibody unconjugated [clone: 11C2]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24928-50EA 782 EUR
ABCAAB24928-50
Anti-EGFR Mouse monoclonal antibody unconjugated [clone: 11C2]
Anticorps
Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: monoclonal
Clone: 11C2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR