Imprimer

Anti-Interferon gamma Mouse Monoclonal Antibody [clone: A151D5B8]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24968-100EA 691 EUR
ABCAAB24968-100
Anti-Interferon gamma Mouse Monoclonal Antibody [clone: A151D5B8]
Anticorps
Type: Primary
Antigen: Interferon gamma
Clonality: monoclonal
Clone: A151D5B8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Pig
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR