Imprimer

Anti-SELL Rat monoclonal antibody Biotin [clone: MEL-14]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB24969-100EA 591 EUR
ABCAAB24969-100
Anti-SELL Rat monoclonal antibody Biotin [clone: MEL-14]
Type: Primary
Antigen: SELL (selectin L)
Clonality: monoclonal
Clone: MEL-14
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR