Imprimer

Anti-Sca1 / Ly6A/E Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: D7]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25031-100EA 746 EUR
ABCAAB25031-100
Anti-Sca1 / Ly6A/E Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: D7]
Anticorps
Type: Primary
Antigen: Sca1 / Ly6A/E
Clonality: monoclonal
Clone: D7
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR