Imprimer

Anti-IL12 p70 Mouse Monoclonal Antibody [clone: QS-12p70]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25036-100EA 698 EUR
ABCAAB25036-100
Anti-IL12 p70 Mouse Monoclonal Antibody [clone: QS-12p70]
Anticorps
Type: Primary
Antigen: IL12 p70
Clonality: monoclonal
Clone: QS-12p70
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR