Imprimer

Anti-IL13 Mouse Monoclonal Antibody [clone: B-B13]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25037-100EA 698 EUR
ABCAAB25037-100
Anti-IL13 Mouse Monoclonal Antibody [clone: B-B13]
Anticorps
Type: Primary
Antigen: IL13 (interleukin 13)
Clonality: monoclonal
Clone: B-B13
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR