Imprimer

Anti-CYCSP51 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25134-100EA 736 EUR
ABCAAB25134-100
Anti-CYCSP51 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: CYCSP51 (cytochrome c, somatic pseudogene 51)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR