Imprimer

Anti-Ly49C+F+H+I Hamster monoclonal antibody [clone: 14B11]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25225-100EA 565 EUR
ABCAAB25225-100
Anti-Ly49C+F+H+I Hamster monoclonal antibody [clone: 14B11]
Type: Primary
Antigen: Ly49C+F+H+I
Clonality: monoclonal
Clone: 14B11
Conjugation: unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR