Imprimer

Anti-CD28 Armenian Hamster Monoclonal Antibody [clone: PV-1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25234-100EA 878 EUR
ABCAAB25234-100
Anti-CD28 Armenian Hamster Monoclonal Antibody [clone: PV-1]
Anticorps
Type: Primary
Antigen: CD28 (CD28 molecule)
Clonality: monoclonal
Clone: PV-1
Conjugation: unconjugated
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR