Imprimer

Anti-ITGB4 Rat Monoclonal Antibody [clone: 346-11A]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25254-100EA 739 EUR
ABCAAB25254-100
Anti-ITGB4 Rat Monoclonal Antibody [clone: 346-11A]
Anticorps
Type: Primary
Antigen: ITGB4 (integrin, beta 4)
Clonality: monoclonal
Clone: 346-11A
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR