Imprimer

Anti-IL4 Rat monoclonal antibody [clone: BVD6-24G2]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25258-100EA 572 EUR
ABCAAB25258-100
Anti-IL4 Rat monoclonal antibody [clone: BVD6-24G2]
Type: Primary
Antigen: IL4 (interleukin 4)
Clonality: monoclonal
Clone: BVD6-24G2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR