Imprimer

Anti-THY1 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: G7]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25272-50EA 698 EUR
ABCAAB25272-50
Anti-THY1 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: G7]
Anticorps
Type: Primary
Antigen: THY1 (Thy-1 cell surface antigen)
Clonality: monoclonal
Clone: G7
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Rat
Isotype: IgG2c
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR