Imprimer

Anti-FAS Mouse monoclonal antibody APC (Allophycocyanin) [clone: DX3]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25291-500EA 319 EUR
ABCAAB25291-500
Anti-FAS Mouse monoclonal antibody APC (Allophycocyanin) [clone: DX3]
Type: Primary
Antigen: FAS (Fas cell surface death receptor)
Clonality: monoclonal
Clone: DX3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR