Imprimer

Anti-TCR alpha + TCR beta Armenian Hamster Monoclonal Antibody [clone: H57-597]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25336-100EA 704 EUR
ABCAAB25336-100
Anti-TCR alpha + TCR beta Armenian Hamster Monoclonal Antibody [clone: H57-597]
Anticorps
Type: Primary
Antigen: TCR alpha + TCR beta
Clonality: monoclonal
Clone: H57-597
Conjugation: unconjugated
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR