Imprimer

Anti-CD4 Rat monoclonal antibody PE (Phycoerythrin)/Cy5® [clone: GK1.5]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25476-50EA 619 EUR
ABCAAB25476-50
Anti-CD4 Rat monoclonal antibody PE (Phycoerythrin)/Cy5® [clone: GK1.5]
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone: GK1.5
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR