Imprimer

Anti-CD19 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 6D5]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25509-50EA 698 EUR
ABCAAB25509-50
Anti-CD19 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 6D5]
Anticorps
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: monoclonal
Clone: 6D5
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR