Imprimer

Anti-Ly6c Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: HK1.4]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25572-100EA 714 EUR
ABCAAB25572-100
Anti-Ly6c Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: HK1.4]
Anticorps
Type: Primary
Antigen: Ly6c
Clonality: monoclonal
Clone: HK1.4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2c
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR