Imprimer

Anti-MS4A1 Mouse Monoclonal Antibody [clone: B-Ly1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25607-100EA 697 EUR
ABCAAB25607-100
Anti-MS4A1 Mouse Monoclonal Antibody [clone: B-Ly1]
Anticorps
Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality: monoclonal
Clone: B-Ly1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR