Imprimer

Anti-CD79B Mouse monoclonal antibody FITC (Fluorescein) [clone: CB3-1]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25626-500EA 241 EUR
ABCAAB25626-500
Anti-CD79B Mouse monoclonal antibody FITC (Fluorescein) [clone: CB3-1]
Type: Primary
Antigen: CD79B (CD79b molecule, immunoglobulin-associated beta)
Clonality: monoclonal
Clone: CB3-1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR