Imprimer

Anti-CCL2 Mouse monoclonal antibody [clone: S8]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25803-100EA 572 EUR
ABCAAB25803-100
Anti-CCL2 Mouse monoclonal antibody [clone: S8]
Type: Primary
Antigen: CCL2 (chemokine (C-C motif) ligand 2)
Clonality: monoclonal
Clone: S8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR