Imprimer

Anti-SIGIRR Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25875-100EA 704 EUR
ABCAAB25875-100
Anti-SIGIRR Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: SIGIRR (single immunoglobulin and toll-interleukin 1 receptor (TIR) domain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR