Imprimer

Anti-West Nile Virus M glycoprotein Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25887-100EA 565 EUR
ABCAAB25887-100
Anti-West Nile Virus M glycoprotein Rabbit polyclonal antibody
Type: Primary
Antigen: West Nile Virus M glycoprotein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR