Imprimer

Anti-KLRB1 Mouse monoclonal antibody PE (Phycoerythrin) [clone: PK136]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB25905-500EA 319 EUR
ABCAAB25905-500
Anti-KLRB1 Mouse monoclonal antibody PE (Phycoerythrin) [clone: PK136]
Type: Primary
Antigen: KLRB1 (killer cell lectin-like receptor subfamily B, member 1)
Clonality: monoclonal
Clone: PK136
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR