Imprimer

Anti-HLA G Mouse Monoclonal Antibody (Biotin) [clone: MEM-G/1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB26034-100EA 705 EUR
ABCAAB26034-100
Anti-HLA G Mouse Monoclonal Antibody (Biotin) [clone: MEM-G/1]
Anticorps
Type: Primary
Antigen: HLA G
Clonality: monoclonal
Clone: MEM-G/1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR