Imprimer

Anti-Laminin Mouse monoclonal antibody FITC (Fluorescein) [clone: 2D11]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB26147-50EA 572 EUR
ABCAAB26147-50
Anti-Laminin Mouse monoclonal antibody FITC (Fluorescein) [clone: 2D11]
Type: Primary
Antigen: Laminin
Clonality: monoclonal
Clone: 2D11
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR