Imprimer

Anti-BCL2L1 Mouse monoclonal antibody FITC (Fluorescein) [clone: 7B2.5]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB26148-50EA 626 EUR
ABCAAB26148-50
Anti-BCL2L1 Mouse monoclonal antibody FITC (Fluorescein) [clone: 7B2.5]
Type: Primary
Antigen: BCL2L1 (BCL2-like 1)
Clonality: monoclonal
Clone: 7B2.5
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR