Imprimer

Anti-SSRP1 Mouse Monoclonal Antibody [clone: 3E4]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB26229-100EA 704 EUR
ABCAAB26229-100
Anti-SSRP1 Mouse Monoclonal Antibody [clone: 3E4]
Anticorps
Type: Primary
Antigen: SSRP1 (structure specific recognition protein 1)
Clonality: monoclonal
Clone: 30000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR