Imprimer

Anti-SFTPD Mouse Monoclonal Antibody [clone: 10B2]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB26267-100EA 691 EUR
ABCAAB26267-100
Anti-SFTPD Mouse Monoclonal Antibody [clone: 10B2]
Anticorps
Type: Primary
Antigen: SFTPD (surfactant protein D)
Clonality: monoclonal
Clone: 10B2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR