Imprimer

Anti-Lysine Rabbit Polyclonal Antibody (Biotin)

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB26332-100EA 691 EUR
ABCAAB26332-100
Anti-Lysine Rabbit Polyclonal Antibody (Biotin)
Anticorps
Type: Primary
Antigen: Lysine
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR