Imprimer

Anti-TSH Mouse monoclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB26403-7EA 565 EUR
ABCAAB26403-7
Anti-TSH Mouse monoclonal antibody
Type: Primary
Antigen: TSH (Thyroid Stimulating Hormone)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR