Imprimer

Anti-CASP14 Rabbit polyclonal antibody unconjugated

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB26991-100EA 691 EUR
ABCAAB26991-100
Anti-CASP14 Rabbit polyclonal antibody unconjugated
Anticorps
Type: Primary
Antigen: CASP14 (caspase 14, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR