Imprimer

Anti-HTLV 1 Tax Mouse Monoclonal Antibody [clone: 1A3]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB26997-50EA 698 EUR
ABCAAB26997-50
Anti-HTLV 1 Tax Mouse Monoclonal Antibody [clone: 1A3]
Anticorps
Type: Primary
Antigen: HTLV 1 Tax
Clonality: monoclonal
Clone: 1A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR