Imprimer

Anti-TUBG1 Mouse Monoclonal Antibody [clone: TU-30]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB27074-100EA 750 EUR
ABCAAB27074-100
Anti-TUBG1 Mouse Monoclonal Antibody [clone: TU-30]
Anticorps
Type: Primary
Antigen: TUBG1 (tubulin, gamma 1)
Clonality: monoclonal
Clone: TU-30
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR