Imprimer

Anti-Cyanazine Mouse monoclonal antibody [clone: HYB269-01]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB27149-100EA 565 EUR
ABCAAB27149-100
Anti-Cyanazine Mouse monoclonal antibody [clone: HYB269-01]
Type: Primary
Antigen: Cyanazine
Clonality: monoclonal
Clone: HYB269-01
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR