Imprimer

Anti-Hydroxytriazine Mouse monoclonal antibody [clone: HYB283-02]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB27151-100EA 565 EUR
ABCAAB27151-100
Anti-Hydroxytriazine Mouse monoclonal antibody [clone: HYB283-02]
Type: Primary
Antigen: Hydroxytriazine
Clonality: monoclonal
Clone: HYB283-02
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR