Imprimer

Anti-Ionotropic glutamate receptor 2 + 3 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB27225-500EA 930 EUR
ABCAAB27225-500
Anti-Ionotropic glutamate receptor 2 + 3 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Ionotropic glutamate receptor 2 + 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR