Imprimer

Anti-IgG Goat Polyclonal Antibody (Gold, 10nm)

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB27241-500EA 518 EUR
ABCAAB27241-500
Anti-IgG Goat Polyclonal Antibody (Gold, 10nm)
Anticorps
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: 10 nm Gold
Epitope:
Host: Goat
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR