Imprimer

Anti-ICAM1 Mouse monoclonal antibody FITC (Fluorescein) [clone: B-H17]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB27297-1EA 241 EUR
ABCAAB27297-1
Anti-ICAM1 Mouse monoclonal antibody FITC (Fluorescein) [clone: B-H17]
Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: monoclonal
Clone: B-H17
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR