Imprimer

Anti-IL2RA Mouse Monoclonal Antibody [clone: B-G3]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB27308-100EA 843 EUR
ABCAAB27308-100
Anti-IL2RA Mouse Monoclonal Antibody [clone: B-G3]
Anticorps
Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: monoclonal
Clone: B-G3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR